1-Phenylimidazole
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002610-5G
002610-1G
FLUO002610-5GEA
0
GBP
FLUO002610-5G
FLUO002610-1G
1-Phenylimidazole
1-Phenylimidazole
Formula:
C₉H₈N₂ Boiling Pt: 276 °C (1013 hPa) Melting Pt: 13 °C Density: 1.1397 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00041204 CAS Number: 7164-98-9 UN: 2384 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...