Dimethyl sulphide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: DMS

M-Clarity™ quality level = MQ300

W274615-1KG-K W274615-25KG-K W274615-5KG-K W274615-9KG-K
SIALW274615-1KG-KEA 135 GBP
SIALW274615-1KG-K SIALW274615-25KG-K SIALW274615-5KG-K SIALW274615-9KG-K
Dimethyl sulphide, Sigma-Aldrich®
Dimethyl sulphide
Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 36.2
Melting Pt: –98 °C
Density: 0.85 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008562
CAS Number: 75-18-3
EINECS: 200-846-2
UN: 1164
ADR: 3 II
Merck Index: 12,06204

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR