Dimethyl disulphide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Dimethyl disulphide, Methyl disulphide

M-Clarity™ quality level = MQ100

471569-1L 471569-250ML 471569-25ML
SIAL471569-1LEA 102 GBP
SIAL471569-1L SIAL471569-250ML SIAL471569-25ML
Dimethyl disulphide, Sigma-Aldrich®
Dimethyl disulphide
Formula: C₂H₆S₂
MW: 94.2 g/mol
Boiling Pt: 110
Melting Pt: –98 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 16
Storage Temperature: Ambient
MDL Number: MFCD00008561
CAS Number: 624-92-0
EINECS: 210-871-0
UN: 2381
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR