Dibutyl sulphide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Dibutylsulphide, Butyl sulphide, n-Butyl sulphide
51491-100ML
51491-500ML
SIAL51491-100MLEA
103
GBP
SIAL51491-100ML
SIAL51491-500ML
Dibutyl sulphide, Sigma-Aldrich®
Dibutyl sulphide
Formula:
C₈H₁₈S Boiling Pt: 189 °C (1013 hPa) Melting Pt: –80 °C Density: 0.83 g/cm³ (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00009468 CAS Number: 544-40-1 EINECS: 208-870-5 UN: 3334 ADR: 9, Merck Index: 12,01626 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...