Selenium disulphide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Selenium sulphide, Selenium(IV) sulphide
M-Clarity™ quality level = MQ100
257710-100G
257710-25G
SIAL257710-100GEA
539
GBP
SIAL257710-100G
SIAL257710-25G
Selenium disulphide, Sigma-Aldrich®
Selenium disulphide
Formula:
S₂Se MW: 143.09 g/mol Boiling Pt: –112 °C (1013 hPa) Density: 0.03 g/cm³ (–65 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011227 CAS Number: 7488-56-4 EINECS: 231-303-8 UN: 2657 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...