Ammonium sulphate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

M-Clarity™ quality level = MQ200

A4418-100G A4418-1KG A4418-500G A4418-5KG
SIALA4418-100GEA 34.2 GBP
SIALA4418-100G SIALA4418-1KG SIALA4418-500G SIALA4418-5KG
Ammonium sulphate, Sigma-Aldrich®
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR