Tetraphenylphosphonium bromide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
218782-100G
218782-25G
SIAL218782-100GEA
325
GBP
SIAL218782-100G
SIAL218782-25G
Tetraphenylphosphonium bromide, Sigma-Aldrich®
Tetraphenylphosphonium bromide
Formula:
C₂₄H₂₀BrP MW: 419.3 g/mol Melting Pt: 299…300 °C |
MDL Number:
MFCD00011915 CAS Number: 2751-90-8 EINECS: 220-393-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...