Tetraphenylphosphonium bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009348-10G 009348-50G 009348-250G
FLUO009348-10GEA 0 GBP
FLUO009348-10G FLUO009348-50G FLUO009348-250G
Tetraphenylphosphonium bromide
Tetraphenylphosphonium bromide
Formula: C₂₄H₂₀BrP
Melting Pt: 299…300 °C
MDL Number: MFCD00011915
CAS Number: 2751-90-8
EINECS: 220-393-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR