Triphenylarsine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
T81906-100G
T81906-25G
T81906-5G
SIALT81906-100GEA
489
GBP
SIALT81906-100G
SIALT81906-25G
SIALT81906-5G
Triphenylarsine, Sigma-Aldrich®
Triphenylarsine
Formula:
C₁₈H₁₅As Boiling Pt: ∼360 °C (1013 hPa) Melting Pt: 58 °C Density: 1.306 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002994 CAS Number: 603-32-7 EINECS: 210-032-9 UN: 3465 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...