Nitronium hexafluoroantimonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

006889-5G 006889-25G
FLUO006889-5GEA 0 GBP
FLUO006889-5G FLUO006889-25G
Nitronium hexafluoroantimonate
Nitronium hexafluoroantimonate
Formula: F₆NO₂Sb
Storage Temperature: Refrigerator
MDL Number: MFCD00042551
CAS Number: 17856-92-7
UN: 1549
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR