Tetrabutylammonium iodide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Tetra-n-butylammonium iodide, TBAI
M-Clarity™ quality level = MQ200
140775-100G
140775-25G
140775-500G
SIAL140775-100GEA
74.2
GBP
SIAL140775-100G
SIAL140775-25G
SIAL140775-500G
Tetrabutylammonium iodide, Sigma-Aldrich®
Tetrabutylammonium iodide
Formula:
C₁₆H₃₆IN MW: 369.37 g/mol Melting Pt: 143…146 °C Density: 1.0027 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011636 CAS Number: 311-28-4 EINECS: 206-220-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...