Tetrabutylammonium fluoride trihydrate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007313-100G 007313-25G
FLUO007313-100GEA 0 GBP
FLUO007313-100G FLUO007313-25G
Tetrabutylammonium fluoride trihydrate
Tetrabutylammonium fluoride trihydrate
Formula: C₁₆H₃₆FN·3H₂O
Melting Pt: 62…63 °C
MDL Number: MFCD00149981
CAS Number: 87749-50-6
EINECS: 207-057-2
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR