Tetramethylammonium bromide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
195758-100G
195758-500G
SIAL195758-100GEA
82.8
GBP
SIAL195758-100G
SIAL195758-500G
Tetramethylammonium bromide, Sigma-Aldrich®
Tetramethylammonium bromide
Formula:
C₄H₁₂BrN Boiling Pt: ∼360 °C (1013 hPa) Density: 1.56 g/cm³ (20 °C) |
MDL Number:
MFCD00011626 CAS Number: 64-20-0 EINECS: 200-581-2 UN: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...