Tetramethylammonium tetrafluoroborate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Tetramethylammoniumtetrafluoroborate

007410-25G 007410-100G 007410-250G
FLUO007410-25GEA 0 GBP
FLUO007410-25G FLUO007410-100G FLUO007410-250G
Tetramethylammonium tetrafluoroborate
Tetramethylammonium tetrafluoroborate
Formula: C₄H₁₂BF₄N
Melting Pt: 418 °C
MDL Number: MFCD00011745
CAS Number: 661-36-9
EINECS: 211-547-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR