Tributylphosphine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Tri-n-butylphosphine, TBP
M-Clarity™ quality level = MQ200
247049-100G
247049-500G
SIAL247049-100GEA
135
GBP
SIAL247049-100G
SIAL247049-500G
Tributylphosphine, Sigma-Aldrich®
Tributylphosphine
Formula:
C₁₂H₂₇P MW: 202.32 g/mol Boiling Pt: 240 °C (1013 hPa) Melting Pt: –65 °C Density: 0.8165 g/cm³ (20 °C) Flash Pt: 40 °C |
MDL Number:
MFCD00009462 CAS Number: 998-40-3 EINECS: 213-651-2 UN: 3254 ADR: 4.2,I |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...