(Isopentyl)triphenylphosphonium bromide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (Isoamyl)triphenylphosphonium bromide, Isopentyltriphenylphosphonium bromide

008045-25G 008045-100G
FLUO008045-25GEA 0 GBP
FLUO008045-25G FLUO008045-100G
(Isopentyl)triphenylphosphonium bromide
(Isopentyl)triphenylphosphonium bromide
Formula: (H₃C)₂HCCH₂CH₂P(C₆H₅)₃Br
MDL Number: MFCD00031594
CAS Number: 28322-40-9
EINECS: 248-966-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR