Bismuth(V) fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004112-10G 004112-25G
FLUO004112-10GEA 0 GBP
FLUO004112-10G FLUO004112-25G
Bismuth(V) fluoride
Bismuth (V) fluoride
Formula: BiF₅
Boiling Pt: 230 °C (1013 hPa)
Melting Pt: 154 °C
Density: 5.55 g/cm³ (20 °C)
MDL Number: MFCD00015967
CAS Number: 7787-62-4
EINECS: 232-125-3
UN: 3084
ADR: 8,I
Merck Index: 12,01315

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR