Diphenyltin oxide
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Diphenyltin(IV) oxide, Diphenyloxostannane
39615.06
39615.14
39615.06EA
0
GBP
39615.06
39615.14
Diphenyltin oxide
Diphenyltin oxide
CAS No.: 2273-51-0
Molecular Formula: (C6H5)2SnO
Formula Weight: 288.90
Physical Form: Powder
Melting Point: 320° dec.
UN No.: 3146
MDL No.: MFCD00058851
Formula:
C₁₂H₁₀OSn Melting Pt: 320 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00058851 CAS Number: 2273-51-0 EINECS: 218-883-8 UN: 3146 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...