Chlorotriethylsilane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

235067-25G 235067-5G
SIAL235067-25GEA 66.5 GBP
SIAL235067-25G SIAL235067-5G
Chlorotriethylsilane, Sigma-Aldrich®
Chlorotriethylsilane
Formula: C₆H₁₅ClSi
Boiling Pt: 144 °C (1013 hPa)
Density: 0.9 g/cm³ (20 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2986
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR