Bromotrimethylsilane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Trimethylbromosilane, Trimethylsilyl bromide
M-Clarity™ quality level = MQ200
194409-100G
194409-25G
194409-5G
SIAL194409-100GEA
236
GBP
SIAL194409-100G
SIAL194409-25G
SIAL194409-5G
Bromotrimethylsilane, Sigma-Aldrich®
Bromotrimethylsilane
Formula:
C₃H₉BrSi Boiling Pt: 79 °C (1013 hPa) Melting Pt: –43 °C Density: 1.1895 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000048 CAS Number: 2857-97-8 EINECS: 220-672-0 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...