Dibutyltin oxide ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Dibutyltin(IV) oxide, Dibutyloxostannane, Di-n-butyltin oxide, Di-n-butyltin(IV) oxide
L14491.22
L14491.36
L14491.22EA
25.5
GBP
L14491.22
L14491.36
Dibutyltin oxide ≥98%
Dibutyltin oxide
Purity: 98%
CAS No.: 818-08-6
Molecular Formula: C8H18OSn
Formula Weight: 248.94
Odour: Odourless
Melting Point:
Formula:
C₈H₁₈OSn MW: 248.94 g/mol Melting Pt: >300 °C Density: 1.58 Storage Temperature: Ambient |
MDL Number:
MFCD00001992 CAS Number: 818-08-6 EINECS: 212-449-1 UN: 3146 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...