Dichlorodimethylgermane ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Dimethylgermanium dichloride
34107.04
34107.09
34107.04EA
92.9
GBP
34107.04
34107.09
Dichlorodimethylgermane ≥97%
Dichlorodimethylgermane
Purity: 98+%
CAS No.: 1529-48-2
Molecular Formula: (CH3)2GeCl2
Formula Weight: 173.57
Flash Point: 32°(90°F)
Physical Form: Liquid
Density: 1.51
Refractive Index: 1.460
Sensitivity: Moisture Sensitive
Melting Point: -22°
Boiling Point: 124°
UN No.: 2924
MDL No.: MFCD00013588
Formula:
C₂H₆Cl₂Ge Boiling Pt: 124 °C Melting Pt: –22 °C Density: 1.51 Flash Pt: 32 °C (90 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013588 CAS Number: 1529-48-2 EINECS: 216-216-5 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...