Diethyltin dichloride

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

71155.02 71155.04 71155.09
71155.02EA 34.3 GBP
71155.02 71155.04 71155.09
Diethyltin dichloride
Diethyltin dichloride

CAS No.: 866-55-7 Molecular Formula: C4H10Cl2Sn Formula Weight: 247.72 Physical Form: Crystalline Sensitivity: Moisture Sensitive Appearance: White Melting Point: 84° Boiling Point: 220° MDL No.: MFCD00013609

Formula: C₄H₁₀Cl₂Sn
MW: 247.74 g/mol
Boiling Pt: 220 °C
Melting Pt: 84 °C
Storage Temperature: Ambient
MDL Number: MFCD00013609
CAS Number: 866-55-7
EINECS: 212-750-8
UN: 3146
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR