Vinyltrimethoxysilane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Trimethoxy(vinyl)silane
M-Clarity™ quality level = MQ200
235768-100ML
235768-500ML
SIAL235768-100MLEA
125
GBP
SIAL235768-100ML
SIAL235768-500ML
Vinyltrimethoxysilane, Sigma-Aldrich®
Vinyltrimethoxysilane
Formula:
C₅H₁₂O₃Si MW: 148.23 g/mol Boiling Pt: 123 °C (1013 hPa) Melting Pt: 0 °C Density: 0.97 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008605 CAS Number: 2768-02-7 EINECS: 220-449-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...