Tin(IV) oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

36302.14 36302.22
36302.14EA 91.1 GBP
36302.14 36302.22
Tin(IV) oxide ≥99.9% (metals basis)
Tin (IV) oxide

Purity: 99.9% CAS No.: 18282-10-5 Molecular Formula: O2Sn Formula Weight: 150.69 Physical Form: 3-12mm Sintered Lump Density: 6.95 Refractive Index: 1.9968 Appearance: White Odour: Odourless Melting Point: 1630° Boiling Point: 1800-1900° subl. Merck Reference: 14,8778 MDL No.: MFCD00011244

Formula: O₂Sn
MW: 150.71 g/mol
Boiling Pt: 1800…1900 °C (subl.)
Melting Pt: 1630 °C
Density: 6.95
Storage Temperature: Ambient
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Merck Index: 14,8778

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR