Silicon dioxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide

M-Clarity™ quality level = MQ100

204358-20G 204358-100G 204358-5G
SIAL204358-20GEA 212 GBP
SIAL204358-20G SIAL204358-100G SIAL204358-5G
Silicon dioxide, Sigma-Aldrich®
Silicium dioxide
Formula: SiO₂
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1719 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4
Merck Index: 14,08493

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR