Germanium(IV) oxide, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

483001-10G 483001-50G
SIAL483001-10GEA 280 GBP
SIAL483001-10G SIAL483001-50G
Germanium(IV) oxide, Sigma-Aldrich®
Germanium (IV) oxide
Formula: GeO₂
Melting Pt: 1110 °C
Density: 3.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011030
CAS Number: 1310-53-8
EINECS: 215-180-8
Merck Index: 12,04417

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR