Trichlorosilane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
175552-100G
175552-500G
175552-5G
SIAL175552-100GEA
108
GBP
SIAL175552-100G
SIAL175552-500G
SIAL175552-5G
Trichlorosilane, Sigma-Aldrich®
Trichlorosilane
Formula:
HCl₃Si Boiling Pt: 32 °C (1013 hPa) Melting Pt: –127.2 °C Density: 1.344 g/cm³ (20 °C) Flash Pt: –28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011518 CAS Number: 10025-78-2 EINECS: 233-042-5 UN: 1295 ADR: 4.3,I REACH: 01-2119494046-35 Merck Index: 12,09776 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...