Lead(II) chloride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ200

203572-10G 203572-50G
SIAL203572-10GEA 159 GBP
SIAL203572-10G SIAL203572-50G
Lead(II) chloride, Sigma-Aldrich®
Lead (II) chloride
Formula: Cl₂Pb
Boiling Pt: 950 °C (1013 hPa)
Melting Pt: 496 °C
Density: 5.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011157
CAS Number: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1,III
Merck Index: 13,05422

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR