Tetrabromosilane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

494100-25ML 494100-5ML
SIAL494100-25MLEA 596 GBP
SIAL494100-25ML SIAL494100-5ML
Tetrabromosilane, Sigma-Aldrich®
Tetrabromosilane
Formula: Br₄Si
Boiling Pt: 154 °C (1013 hPa)
Melting Pt: 5 °C
Density: 2.812 g/cm³ (25 °C)
MDL Number: MFCD00049530
CAS Number: 7789-66-4
EINECS: 232-182-4
UN: 3264
ADR: 8,II
Merck Index: 12,08643

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR