Thallium(I) sulphate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Thallic sulphate, Dithallium sulphate
M-Clarity™ quality level = MQ200
208191-250G
208191-50G
SIAL208191-250GEA
998
GBP
SIAL208191-250G
SIAL208191-50G
Thallium(I) sulphate, Sigma-Aldrich®
Dithallium sulphate
Formula:
O₄STl₂ Melting Pt: 632 °C Density: 6.77 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011278 CAS Number: 7446-18-6 EINECS: 231-201-3 UN: 1707 ADR: 6.1,II Merck Index: 12,09405 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...