Orthoboric acid
Supplier: Roth Carl
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
5935.1
5935.2
5935.3
ROTH5935.1EA
0
GBP
ROTH5935.1
ROTH5935.2
ROTH5935.3
Orthoboric acid
ortho-Boric acid
Formula:
B(OH)₃ MW: 61.83 g/mol Boiling Pt: 185 Melting Pt: 160 Density: 1.44 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011337 CAS Number: 10043-35-3 EINECS: 233-139-2 REACH: 01-2119486683-25 Merck Index: 13,01326 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...