Aluminium nitride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Aluminum nitride

M-Clarity™ quality level = MQ100

593044-10G 593044-50G
SIAL593044-10GEA 176 GBP
SIAL593044-10G SIAL593044-50G
Aluminium nitride, Sigma-Aldrich®
Aluminium nitride
Formula: AlN
Boiling Pt: 2517 °C (1013 hPa)
Melting Pt: 1900…2200 °C
Density: 3.257…3.267 g/cm³ (160 °C)
MDL Number: MFCD00003429
CAS Number: 24304-00-5
EINECS: 246-140-8
Merck Index: 12,00366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR