Aluminium nitride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Aluminum nitride
241903-250G
241903-50G
SIAL241903-250GEA
359
GBP
SIAL241903-250G
SIAL241903-50G
Aluminium nitride, Sigma-Aldrich®
Aluminium nitride
Formula:
AlN MW: 40.99 g/mol Boiling Pt: 2517 °C (1013 hPa) Melting Pt: 1900…2200 °C Density: 3.257…3.267 g/cm³ (160 °C) |
MDL Number:
MFCD00003429 CAS Number: 24304-00-5 EINECS: 246-140-8 Merck Index: 12,00366 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...