Magnesium fluoride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
378836-25G
378836-5G
SIAL378836-25GEA
258
GBP
SIAL378836-25G
SIAL378836-5G
Magnesium fluoride, Sigma-Aldrich®
Magnesium fluoride
Formula:
MgF₂ Boiling Pt: 2260 °C (1013 hPa) Melting Pt: 1266 °C Density: 3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011108 CAS Number: 7783-40-6 EINECS: 231-995-1 Merck Index: 12,05701 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...