Magnesium fluoride, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

343188-250G 343188-25G
SIAL343188-250GEA 205 GBP
SIAL343188-250G SIAL343188-25G
Magnesium fluoride, Sigma-Aldrich®
Magnesium fluoride
Formula: MgF₂
Boiling Pt: 2260 °C (1013 hPa)
Melting Pt: 1266 °C
Density: 3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011108
CAS Number: 7783-40-6
EINECS: 231-995-1
Merck Index: 12,05701

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR