Strontium carbonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
472018-100G
472018-500G
SIAL472018-100GEA
67.3
GBP
SIAL472018-100G
SIAL472018-500G
Strontium carbonate, Sigma-Aldrich®
Strontium carbonate
Formula:
CO₃Sr Boiling Pt: 2647 °C (1 mmHg) Melting Pt: 1768.85 °C Density: 3.548 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011250 CAS Number: 1633-05-2 EINECS: 216-643-7 Merck Index: 14,08838 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...