Calcium carbonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Marble
M-Clarity™ quality level = MQ100
481807-25G
481807-5G
SIAL481807-25GEA
972
GBP
SIAL481807-25G
SIAL481807-5G
Calcium carbonate, Sigma-Aldrich®
Calcium carbonate
Formula:
CaCO₃ Melting Pt: 825 °C Density: 2.71 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010906 CAS Number: 471-34-1 EINECS: 207-439-9 Merck Index: 13,01658 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...