Ammonium hydrogen difluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

004041-5KG 004041-10KG 004041-1KG
FLUO004041-5KGEA 0 GBP
FLUO004041-5KG FLUO004041-10KG FLUO004041-1KG
Ammonium hydrogen difluoride
Ammonium hydrogen difluoride
Formula: NH₄HF₂
Melting Pt: 127 °C
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8,II
Merck Index: 13,00498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR