Sodium selenate, BioXtra, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
S8295-10G
S8295-25G
SIALS8295-10GEA
82.9
GBP
SIALS8295-10G
SIALS8295-25G
Sodium selenate, BioXtra, Sigma-Aldrich®
Sodium selenate
Formula:
Na₂O₄Se MW: 188.94 g/mol Density: 3.098 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003490 CAS Number: 13410-01-0 EINECS: 236-501-8 UN: 2630 ADR: 6.1,I Merck Index: 12,08820 |
Specification Test Results
Appearance (Color) | White to Off White |
Appearance (Form) | Powder |
Insoluble matter | ≤ 0.1 % |
Chloride Content | ≤ 0.05 % |
Aluminum (Al) | ≤ 0.0005 % |
Phosphorus (P) | ≤ 0.01 % |
Lead (Pb) | ≤ 0.001 % |
Iron (Fe) | ≤ 0.0005 % |
Copper (Cu) | ≤ 0.0005 % |
Zinc (Zn) | ≤ 0.0005 % |
Calcium (Ca) | ≤ 0.005 % |
Magnesium (Mg) | ≤ 0.0005 % |
Potassium (K) | ≤ 0.02 % |
Ammonia (NH4) | ≤ 0.05 % |
Sulfate (SO4) | ≤ 0.1 % |
Selenium (Se) | > 41 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...