Sodium dihydrogen phosphate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic
M-Clarity™ quality level = MQ200
S3139-1KG
S3139-250G
S3139-500G
SIALS3139-1KGEA
489
GBP
SIALS3139-1KG
SIALS3139-250G
SIALS3139-500G
Sodium dihydrogen phosphate, Sigma-Aldrich®
Sodium dihydrogen phosphate
Formula:
NaH₂PO₄ Melting Pt: ∼650 °C Density: 0.9996 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003527 CAS Number: 7558-80-7 EINECS: 231-449-2 REACH: 01-2119489796-13 Merck Index: 12,08806 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...