Potassium hexafluorophosphate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
002859-250G
002859-1KG
FLUO002859-250GEA
0
GBP
FLUO002859-250G
FLUO002859-1KG
Potassium hexafluorophosphate
Potassium hexafluorophosphate
Formula:
F₆KP Melting Pt: 575 °C Density: 2.58 g/cm³ (20 °C) |
MDL Number:
MFCD00011412 CAS Number: 17084-13-8 EINECS: 241-143-0 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...