Rubidium nitrate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger


M-Clarity™ quality level = MQ100

289299-10G 289299-50G
SIAL289299-10GEA 56.3 GBP
SIAL289299-10G SIAL289299-50G
Rubidium nitrate, Sigma-Aldrich®
Rubidium nitrate
Formula: NO₃Rb
MW: 147.47 g/mol
Boiling Pt: 578
Melting Pt: 310
Density: 3.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011193
CAS Number: 13126-12-0
EINECS: 236-060-1
UN: 1477
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR