Rubidium fluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007252-10G 007252-25G 007252-100G
FLUO007252-10GEA 0 GBP
FLUO007252-10G FLUO007252-25G FLUO007252-100G
Rubidium fluoride
Rubidium fluoride
Formula: FRb
MW: 104.47 g/mol
Boiling Pt: 1408
Melting Pt: 753
Density: 3.983 g/cm³ (–273.15 °C)
MDL Number: MFCD00011191
CAS Number: 13446-74-7
EINECS: 236-603-2
UN: 3288
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR