Sodium fluoride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ200
S7920-500G
S7920-100G
SIALS7920-500GEA
455
GBP
SIALS7920-500G
SIALS7920-100G
Sodium fluoride, Sigma-Aldrich®
Sodium fluoride
Formula:
NaF Boiling Pt: 1700 °C (1013 hPa) Melting Pt: 993 °C Density: 2.8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003524 CAS Number: 7681-49-4 EINECS: 231-667-8 UN: 1690 ADR: 6.1,III Merck Index: 13,08691 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...