Sodium carbonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Calcined soda, Carbonic acid disodium salt
M-Clarity™ quality level = MQ200
S7795-1KG
S7795-500G
SIALS7795-1KGEA
180
GBP
SIALS7795-1KG
SIALS7795-500G
Sodium carbonate, Sigma-Aldrich®
Sodium carbonate
Formula:
Na₂CO₃ MW: 105.99 g/mol Boiling Pt: 1600 Melting Pt: 854 Density: 2.53 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 REACH: 01-2119485498-19 Merck Index: 14,08596 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...