Cesium carbonate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
441902-250G
441902-50G
SIAL441902-250GEA
235
GBP
SIAL441902-250G
SIAL441902-50G
Cesium carbonate, Sigma-Aldrich®
Cesium carbonate
Formula:
Cs₂CO₃ MW: 325.82 g/mol Melting Pt: 610 °C Density: 4.24 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010957 CAS Number: 534-17-8 EINECS: 208-591-9 Merck Index: 13,02020 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...