Cesium carbonate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ200

202126-100G 202126-25G 202126-500G 202126-5G
SIAL202126-100GEA 182 GBP
SIAL202126-100G SIAL202126-25G SIAL202126-500G SIAL202126-5G
Cesium carbonate, Sigma-Aldrich®
Cesium carbonate
Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: 610
Density: 4.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 13,02020

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR