Cesium chloride
Supplier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Cesium Chloride
15554.02
15554.03
15554.04
SERA15554.02EA
0
GBP
SERA15554.02
SERA15554.03
SERA15554.04
Cesium chloride
Cesium chloride
Formula:
CsCl MW: 168.36 g/mol Boiling Pt: 1303 Melting Pt: 646 Density: 3.7808 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010955 CAS Number: 7647-17-8 EINECS: 231-600-2 Merck Index: 13,02021 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...