Potassium tetrahydroborate, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
M-Clarity™ quality level = MQ100
60080-100G
60080-25G
SIAL60080-100GEA
123
GBP
SIAL60080-100G
SIAL60080-25G
Potassium tetrahydroborate, Sigma-Aldrich®
Potassium tetrahydroborate
Formula:
H₄BK Melting Pt: 500 °C Density: 1.17 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011396 CAS Number: 13762-51-1 EINECS: 237-360-5 UN: 1870 ADR: 4.3,I Merck Index: 12,07777 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...